Product Details

SNP ID
rs1226588
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:100578732 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCTCTGTTAACCAAAGTACTATCT[A/G]TCATTTCGGTATTTTGAGCTTCTCA
Phenotype
MIM: 603421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSANTD3-TMEFF1 PubMed Links

Gene Details

Gene
MSANTD3-TMEFF1
Gene Name
MSANTD3-TMEFF1 readthrough
There are no transcripts associated with this gene.

Gene
MURC
Gene Name
muscle related coiled-coil protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018116.2 Intron NP_001018126.1
Gene
TMEFF1
Gene Name
transmembrane protein with EGF like and two follistatin like domains 1
There are no transcripts associated with this gene.

View Full Product Details