Product Details

SNP ID
rs3814132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124853697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTCCAGAATGTTCTCTGGGCAGT[A/G]GCTCAGGACATTGAGTCCAGAGTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPC5L PubMed Links

Gene Details

Gene
ARPC5L
Gene Name
actin related protein 2/3 complex subunit 5 like
There are no transcripts associated with this gene.

Gene
RPL35
Gene Name
ribosomal protein L35
There are no transcripts associated with this gene.

Gene
WDR38
Gene Name
WD repeat domain 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045476.2 Intron NP_001038941.1
NM_001276374.1 Intron NP_001263303.1
NM_001276375.1 Intron NP_001263304.1
NM_001276376.1 Intron NP_001263305.1
XM_005251987.3 Intron XP_005252044.1

View Full Product Details