Product Details
- SNP ID
-
rs3765975
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
59
- Location
-
Chr.1:86427136 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- TGTCCTTCCAAGTCTACAATGAGAT[G/T]TAGGTAGGAATCATGGAAGGGGGAA
- Phenotype
-
MIM: 604003
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLCA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111331175] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLCA2
- Gene Name
- chloride channel accessory 2
View Full Product Details