Product Details

SNP ID
rs12892769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20427510 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTCTGTTTCAAATGCATTCCCC[C/G]TTCTCTAATGGGTGAGACTCTAGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLHL33 PubMed Links

Gene Details

Gene
KLHL33
Gene Name
kelch like family member 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109997.2 4163 Intron NP_001103467.2
XM_005267347.4 4163 UTR 3 XP_005267404.1
XM_011536450.2 4163 Intron XP_011534752.1

View Full Product Details