Product Details

SNP ID
rs945012
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:20427487 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACAGGGCATGGCATGCCCCACTTC[C/T]TTCTGTTTCAAATGCATTCCCCCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL33 PubMed Links

Gene Details

Gene
KLHL33
Gene Name
kelch like family member 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109997.2 4186 Intron NP_001103467.2
XM_005267347.4 4186 UTR 3 XP_005267404.1
XM_011536450.2 4186 Intron XP_011534752.1

View Full Product Details