Product Details

SNP ID
rs3738772
Assay Type
Validated
NCBI dbSNP Submissions
57
Location
Chr.1:109505814 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
ACTAGAAACGTGGGAGAGAAGAGAC[A/C]CTAAGAAAAACCAACATCAGAGAAC
Phenotype
MIM: 615689
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AMIGO1 PubMed Links

Gene Details

Gene
AMIGO1
Gene Name
adhesion molecule with Ig like domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020703.2 3487 Intron NP_065754.2
XM_011541812.2 3487 UTR 3 XP_011540114.1
Gene
CYB561D1
Gene Name
cytochrome b561 family member D1
There are no transcripts associated with this gene.

View Full Product Details