Product Details
- SNP ID
-
rs10830124
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:127552145 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTAAGTGAATTTCAAAAACAATTC[A/C]CAGGGCTAATTAGTGCTGCTTGAAA
- Phenotype
-
MIM: 609513
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NPS
PubMed Links
Gene Details
- Gene
- NPS
- Gene Name
- neuropeptide S
View Full Product Details