Product Details
- SNP ID
-
rs9449648
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:83514041 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATGGAGAGGTTAGGAAACAGTAAA[A/G]TTAAGTTAGTAGTATGCAAGCTATT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS35
PubMed Links
Gene Details
- Gene
- PRSS35
- Gene Name
- protease, serine 35
View Full Product Details