Product Details

SNP ID
rs6933234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107156862 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTTTGACTGATAACAATGAATCC[C/T]AGTCTTTCTATCTCTTCAGAGCTCT
Phenotype
MIM: 610564
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDSS2 PubMed Links

Gene Details

Gene
PDSS2
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020381.3 Intron NP_065114.3
XM_011535956.2 Intron XP_011534258.1
XM_011535957.2 Intron XP_011534259.1
XM_011535958.2 Intron XP_011534260.1
XM_011535959.2 Intron XP_011534261.1
XM_011535960.2 Intron XP_011534262.1
XM_011535961.2 Intron XP_011534263.1
XM_011535962.2 Intron XP_011534264.1
XM_011535963.2 Intron XP_011534265.1
XM_017011082.1 Intron XP_016866571.1

View Full Product Details