Product Details
- SNP ID
-
rs11245357
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124763473 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAACAAGACCTTTGGCAGTGACGCA[A/G]GGCAGTGTATTCTGTTACTGTCCCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
METTL10
PubMed Links
Gene Details
- Gene
- METTL10
- Gene Name
- methyltransferase like 10
View Full Product Details