Product Details
- SNP ID
-
rs10776724
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
67
- Location
-
Chr.1:111326865 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGTAATGTTGAGTTCTTTTCATAT[A/C]CACGTTAGCCATTTCTTTGTCTTCT
- Phenotype
-
MIM: 606080
MIM: 614234
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CHIA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75599456] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHIA
- Gene Name
- chitinase, acidic
There are no transcripts associated with this gene.
- Gene
- PIFO
- Gene Name
- primary cilia formation
View Full Product Details