Product Details

SNP ID
rs939883
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:196238244 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTCGTGAAGGACTTGGCAAGCCAC[A/T]TAGCTTCAGAACGGAAGGCAAATGC
Phenotype
MIM: 123695 MIM: 612084
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PCYT1A PubMed Links

Gene Details

Gene
PCYT1A
Gene Name
phosphate cytidylyltransferase 1, choline, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312673.1 1696 UTR 3 NP_001299602.1
NM_005017.3 1696 UTR 3 NP_005008.2
Gene
SLC51A
Gene Name
solute carrier family 51 alpha subunit
There are no transcripts associated with this gene.

View Full Product Details