Product Details

SNP ID
rs11578832
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:53208870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACACCTACCAGAATATCTCCAC[A/G]CCACAAGACATGTAACAAGTGCTGC
Phenotype
MIM: 600650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf123 PubMed Links

Gene Details

Gene
C1orf123
Gene Name
chromosome 1 open reading frame 123
There are no transcripts associated with this gene.

Gene
CPT2
Gene Name
carnitine palmitoyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000098.2 Intron NP_000089.1
XM_005270484.1 Intron XP_005270541.1

View Full Product Details