Product Details
- SNP ID
-
rs17075821
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:114056982 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGAATCAAAGGAAGACTAACTCT[C/T]TGAGAATAGCTGAAGTCACTTTCCC
- Phenotype
-
MIM: 609407
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HS3ST5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74668424] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HS3ST5
- Gene Name
- heparan sulfate-glucosamine 3-sulfotransferase 5
- Gene
- LOC101927768
- Gene Name
- uncharacterized LOC101927768
There are no transcripts associated with this gene.
View Full Product Details