Product Details

SNP ID
rs4516634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183644402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGTAGTTACGGAGACCAGTAAT[G/T]GGAGAGTTCCCAGAAAATCCAGAGG
Phenotype
MIM: 611295
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL24 PubMed Links

Gene Details

Gene
KLHL24
Gene Name
kelch like family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017644.3 542 Intron NP_060114.2
XM_005247552.2 542 Intron XP_005247609.1
XM_005247553.1 542 Intron XP_005247610.1
XM_005247554.2 542 Intron XP_005247611.1
XM_005247555.1 542 Intron XP_005247612.1
XM_005247556.1 542 Intron XP_005247613.1
XM_011512938.2 542 Intron XP_011511240.1
XM_017006650.1 542 Intron XP_016862139.1
XM_017006651.1 542 Intron XP_016862140.1
XM_017006652.1 542 Intron XP_016862141.1
XM_017006653.1 542 Intron XP_016862142.1
XM_017006654.1 542 Intron XP_016862143.1
XM_017006655.1 542 Intron XP_016862144.1
XM_017006656.1 542 Intron XP_016862145.1
XM_017006657.1 542 Intron XP_016862146.1
XM_017006658.1 542 Intron XP_016862147.1
XM_017006659.1 542 UTR 5 XP_016862148.1
XM_017006660.1 542 Intron XP_016862149.1
XM_017006661.1 542 Intron XP_016862150.1
XM_017006662.1 542 Intron XP_016862151.1
XM_017006663.1 542 Intron XP_016862152.1

View Full Product Details