Product Details

SNP ID
rs17154155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:80604927 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTCTAATTGAAATTCATTTGGAG[G/T]TTCCTTTGGTCCCATGTTGGAACTT
Phenotype
MIM: 173510
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD36 PubMed Links

Gene Details

Gene
CD36
Gene Name
CD36 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000072.3 Intron NP_000063.2
NM_001001547.2 Intron NP_001001547.1
NM_001001548.2 Intron NP_001001548.1
NM_001127443.1 Intron NP_001120915.1
NM_001127444.1 Intron NP_001120916.1
NM_001289908.1 Intron NP_001276837.1
NM_001289909.1 Intron NP_001276838.1
NM_001289911.1 Intron NP_001276840.1
XM_005250713.1 Intron XP_005250770.1
XM_005250714.1 Intron XP_005250771.1
XM_005250715.4 Intron XP_005250772.1

View Full Product Details