Product Details

SNP ID
rs7158865
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:55789933 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATAAACACAAATACCCATATTAG[C/T]GTTATGTACTTAGAGATTTTTTAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00520 PubMed Links

Gene Details

Gene
LINC00520
Gene Name
long intergenic non-protein coding RNA 520
There are no transcripts associated with this gene.

View Full Product Details