Product Details

SNP ID
rs10803417
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:16067457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTTTCTTTCAGTCCACACCCTA[C/G]TCTCCTGACCTCAAGGTCAGAATTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM131C PubMed Links

Gene Details

Gene
FAM131C
Gene Name
family with sequence similarity 131 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182623.2 Intron NP_872429.2

View Full Product Details