Product Details
- SNP ID
-
rs2819878
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:80141137 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACGCTGCTACTGAAATATCTCCTT[C/T]CATTTTCTTCCTCACTATTCCTACA
- Phenotype
-
MIM: 612857
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLAC9
PubMed Links
Gene Details
- Gene
- PLAC9
- Gene Name
- placenta specific 9
View Full Product Details