Product Details
- SNP ID
-
rs10020210
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:112244042 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATATTCCTATATACCAGCAACAAAT[A/G]CTGCATATTATAAAATTACCTTTTA
- Phenotype
-
MIM: 610851
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AP1AR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72894916] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AP1AR
- Gene Name
- adaptor related protein complex 1 associated regulatory protein
View Full Product Details