Product Details

SNP ID
rs10882275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93593731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAACCCCTGTTTTCTCTGGGGTA[C/T]GGACAAAATGAATTTCACTAGCACG
Phenotype
MIM: 609044 MIM: 180250
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FFAR4 PubMed Links

Gene Details

Gene
FFAR4
Gene Name
free fatty acid receptor 4
There are no transcripts associated with this gene.

Gene
RBP4
Gene Name
retinol binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323517.1 Intron NP_001310446.1
NM_001323518.1 Intron NP_001310447.1
NM_006744.3 Intron NP_006735.2

View Full Product Details