Product Details

SNP ID
rs7744014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:145802682 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGAATTGTATTTAAGAAAATAT[A/G]TATCACATTAGCTCTTGTGTAGAGA
Phenotype
MIM: 609101
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO30 PubMed Links

Gene Details

Gene
FBXO30
Gene Name
F-box protein 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032145.4 Intron NP_115521.3
XM_005267159.3 Intron XP_005267216.1
XM_011536177.2 Intron XP_011534479.1
XM_017011353.1 Intron XP_016866842.1
Gene
LOC100507557
Gene Name
uncharacterized LOC100507557
There are no transcripts associated with this gene.

View Full Product Details