Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001287426.1 | 431 | UTR 5 | NP_001274355.1 | ||
NM_001287428.1 | 431 | UTR 5 | NP_001274357.1 | ||
NM_001287429.1 | 431 | UTR 5 | NP_001274358.1 | ||
NM_001287430.1 | 431 | UTR 5 | NP_001274359.1 | ||
NM_003364.3 | 431 | UTR 5 | NP_003355.1 | ||
XM_005249838.4 | 431 | UTR 5 | XP_005249895.1 | ||
XM_011515512.2 | 431 | UTR 5 | XP_011513814.1 | ||
XM_011515513.2 | 431 | UTR 5 | XP_011513815.1 | ||
XM_011515514.2 | 431 | UTR 5 | XP_011513816.1 | ||
XM_011515515.2 | 431 | UTR 5 | XP_011513817.1 |