Product Details

SNP ID
rs3736546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47988538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTTAAGAAAGAACCAAGAAATT[A/C]AATATATATATTATAAAAAAGTGAT
Phenotype
MIM: 603760
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HUS1 PubMed Links

Gene Details

Gene
HUS1
Gene Name
HUS1 checkpoint clamp component
There are no transcripts associated with this gene.

Gene
SUN3
Gene Name
Sad1 and UNC84 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030019.1 Intron NP_001025190.1
NM_001284350.1 Intron NP_001271279.1
NM_152782.3 Intron NP_689995.3
XM_017011930.1 Intron XP_016867419.1

View Full Product Details