Product Details

SNP ID
rs34337446
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123969913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCCCGTTTTCTTTCAGATCAT[C/G]CATATTTCATGTTTCTCACAATACT
Phenotype
MIM: 611757
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CCDC14 PubMed Links

Gene Details

Gene
CCDC14
Gene Name
coiled-coil domain containing 14
There are no transcripts associated with this gene.

Gene
ROPN1
Gene Name
rhophilin associated tail protein 1
There are no transcripts associated with this gene.

View Full Product Details