Product Details

SNP ID
rs10917403
Assay Type
Functionally tested
NCBI dbSNP Submissions
62
Location
Chr.1:23639214 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTTCATGGTGTCTGCTTAATGTT[A/C]ATCTCTCCAAGCTTCCCACCAGACT
Phenotype
MIM: 607305
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MDS2 PubMed Links

Gene Details

Gene
MDS2
Gene Name
myelodysplastic syndrome 2 translocation associated
There are no transcripts associated with this gene.

View Full Product Details