Product Details
- SNP ID
-
rs11998791
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:36833370 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATAAAGGTTACATAAAATCAATT[A/C]TTCACAAGTAACAGCCACTCATGAA
- Phenotype
-
MIM: 167414
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4475
PubMed Links
Gene Details
- Gene
- MIR4475
- Gene Name
- microRNA 4475
There are no transcripts associated with this gene.
- Gene
- PAX5
- Gene Name
- paired box 5
View Full Product Details