Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001083963.1 | 2236 | UTR 3 | NP_001077432.1 | ||
NM_001083964.1 | 2236 | UTR 3 | NP_001077433.1 | ||
NM_001083965.1 | 2236 | UTR 3 | NP_001077434.1 | ||
NM_006862.3 | 2236 | Intron | NP_006853.2 | ||
XM_005244856.4 | 2236 | UTR 3 | XP_005244913.1 | ||
XM_017000122.1 | 2236 | UTR 3 | XP_016855611.1 | ||
XM_017000123.1 | 2236 | Intron | XP_016855612.1 | ||
XM_017000124.1 | 2236 | UTR 3 | XP_016855613.1 | ||
XM_017000125.1 | 2236 | UTR 3 | XP_016855614.1 | ||
XM_017000126.1 | 2236 | Intron | XP_016855615.1 | ||
XM_017000127.1 | 2236 | Intron | XP_016855616.1 | ||
XM_017000128.1 | 2236 | UTR 3 | XP_016855617.1 | ||
XM_017000129.1 | 2236 | UTR 3 | XP_016855618.1 | ||
XM_017000130.1 | 2236 | Intron | XP_016855619.1 |