Product Details
- SNP ID
-
rs35211759
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:27115131 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAAGGCCTTAAACAATGACTGAT[A/G]GAGATTGGAGGATAAATACCTCAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NEK10
PubMed Links
Gene Details
- Gene
- NEK10
- Gene Name
- NIMA related kinase 10
View Full Product Details