Product Details

SNP ID
rs11871837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81651952 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCCCAGGATGCTGGGCAGGTGC[A/G]TGCCCTGGGGGAGTACGGGGGGGTG
Phenotype
MIM: 180073
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE6G PubMed Links

Gene Details

Gene
PDE6G
Gene Name
phosphodiesterase 6G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002602.3 Intron NP_002593.1
XM_006721942.3 Intron XP_006722005.1
XM_017024734.1 Intron XP_016880223.1
XM_017024735.1 Intron XP_016880224.1
XM_017024736.1 Intron XP_016880225.1
Gene
TSPAN10
Gene Name
tetraspanin 10
There are no transcripts associated with this gene.

View Full Product Details