Product Details

SNP ID
rs13103004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2973931 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTCTCTCACTCTGCCTCCACAC[A/G]GTTTCCTCTGTCTCTTGAGAATTTT
Phenotype
MIM: 137026
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRK4 PubMed Links

Gene Details

Gene
GRK4
Gene Name
G protein-coupled receptor kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004056.1 Intron NP_001004056.1
NM_001004057.1 Intron NP_001004057.1
NM_005307.2 Intron NP_005298.2
NM_182982.2 Intron NP_892027.2
XM_005247962.3 Intron XP_005248019.1
XM_006713880.3 Intron XP_006713943.1
XM_011513447.2 Intron XP_011511749.1
XM_011513448.2 Intron XP_011511750.1
XM_011513449.2 Intron XP_011511751.1
XM_011513450.2 Intron XP_011511752.1
XM_011513451.2 Intron XP_011511753.1
XM_011513452.2 Intron XP_011511754.1
XM_011513453.2 Intron XP_011511755.1
XM_011513454.2 Intron XP_011511756.1
XM_011513455.2 Intron XP_011511757.1
XM_011513456.2 Intron XP_011511758.1
XM_011513457.2 Intron XP_011511759.1
XM_017008052.1 Intron XP_016863541.1
XM_017008053.1 Intron XP_016863542.1
XM_017008054.1 Intron XP_016863543.1
XM_017008055.1 Intron XP_016863544.1
XM_017008056.1 Intron XP_016863545.1
XM_017008057.1 Intron XP_016863546.1
XM_017008058.1 Intron XP_016863547.1
XM_017008059.1 Intron XP_016863548.1
XM_017008060.1 Intron XP_016863549.1
XM_017008061.1 Intron XP_016863550.1
XM_017008062.1 Intron XP_016863551.1
XM_017008063.1 Intron XP_016863552.1
XM_017008064.1 Intron XP_016863553.1
XM_017008065.1 Intron XP_016863554.1
XM_017008066.1 Intron XP_016863555.1

View Full Product Details