Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001001560.2 | 201 | Intron | NP_001001560.1 | ||
NM_001172687.1 | 201 | Intron | NP_001166158.1 | ||
NM_001172688.1 | 201 | UTR 5 | NP_001166159.1 | ||
NM_013365.4 | 201 | Intron | NP_037497.1 | ||
XM_005261517.3 | 201 | Intron | XP_005261574.1 | ||
XM_005261518.1 | 201 | Intron | XP_005261575.1 | ||
XM_005261520.1 | 201 | Intron | XP_005261577.1 | ||
XM_005261522.1 | 201 | Intron | XP_005261579.1 | ||
XM_006724229.1 | 201 | Intron | XP_006724292.1 | ||
XM_011530122.1 | 201 | Intron | XP_011528424.1 | ||
XM_011530123.1 | 201 | Intron | XP_011528425.1 | ||
XM_011530124.1 | 201 | Intron | XP_011528426.1 | ||
XM_017028760.1 | 201 | Intron | XP_016884249.1 | ||
XM_017028761.1 | 201 | Intron | XP_016884250.1 | ||
XM_017028762.1 | 201 | Intron | XP_016884251.1 | ||
XM_017028763.1 | 201 | Intron | XP_016884252.1 | ||
XM_017028764.1 | 201 | Intron | XP_016884253.1 | ||
XM_017028765.1 | 201 | Intron | XP_016884254.1 |