Product Details

SNP ID
rs12564199
Assay Type
Validated
NCBI dbSNP Submissions
36
Location
Chr.1:153976064 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCTGATGCGGATGGTGGAGGTGGT[A/G]GTTGCATGATTACGAATGTATGTTA
Phenotype
MIM: 607138 MIM: 604671 MIM: 602672 MIM: 604740
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links
Additional Information
For this assay, SNP(s) [rs78714663] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
There are no transcripts associated with this gene.

Gene
JTB
Gene Name
jumping translocation breakpoint
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006694.3 Intron NP_006685.1
Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
There are no transcripts associated with this gene.

View Full Product Details