Product Details

SNP ID
rs12687730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50911598 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACATGTGCTACTGTATTTCTTAA[C/T]GAAAGCTACTATCTTAACTGTCTTG
Phenotype
MIM: 300247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMP15 PubMed Links

Gene Details

Gene
BMP15
Gene Name
bone morphogenetic protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005448.2 Intron NP_005439.2

View Full Product Details