Product Details
- SNP ID
-
rs2235488
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1681305 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TGTGAAACCTGCAGTGGGACCTGTT[G/T]CCTTTGTGACCGAGATAGCCGCTGC
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CRAMP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116251976] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRAMP1
- Gene Name
- cramped chromatin regulator homolog 1
There are no transcripts associated with this gene.
- Gene
- HN1L
- Gene Name
- hematological and neurological expressed 1-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144570.2 |
|
Intron |
|
|
NP_653171.1 |
View Full Product Details