Product Details
- SNP ID
-
rs2858027
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:3505206 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGGGCGACCGTGTCCTGGACAGCCC[A/G]GCTCGAGGCTGCTCTTCAGCCCCAG
- Phenotype
-
MIM: 610285
MIM: 104225
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DOK7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112021500] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DOK7
- Gene Name
- docking protein 7
There are no transcripts associated with this gene.
- Gene
- LRPAP1
- Gene Name
- LDL receptor related protein associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002337.3 |
8927 |
UTR 3 |
|
|
NP_002328.1 |
View Full Product Details