Product Details

SNP ID
rs2444842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75758725 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAAGGGTGGAAGGCCGCGGGGGC[A/G]GGGCAGCCTGGCTCTCTCAGACGTG
Phenotype
MIM: 604313 MIM: 147557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALK1 PubMed Links

Gene Details

Gene
GALK1
Gene Name
galactokinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000154.1 Intron NP_000145.1
Gene
ITGB4
Gene Name
integrin subunit beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000213.4 Intron NP_000204.3
NM_001005619.1 Intron NP_001005619.1
NM_001005731.2 Intron NP_001005731.1
NM_001321123.1 Intron NP_001308052.1
XM_005257309.2 Intron XP_005257366.1
XM_005257311.4 Intron XP_005257368.1
XM_006721866.3 Intron XP_006721929.1
XM_006721867.3 Intron XP_006721930.1
XM_006721868.3 Intron XP_006721931.1
XM_006721870.3 Intron XP_006721933.1
XM_011524751.2 Intron XP_011523053.1
XM_011524752.2 Intron XP_011523054.1

View Full Product Details