Product Details

SNP ID
rs786617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:44366019 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTGAGTGTCAAATCTTTCTAG[A/G]TATTTGTAATTCTTCAAATGTTTAT
Phenotype
MIM: 609559 MIM: 609557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMKMT PubMed Links

Gene Details

Gene
CAMKMT
Gene Name
calmodulin-lysine N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024766.4 Intron NP_079042.1
XM_011533111.2 Intron XP_011531413.1
XM_011533113.2 Intron XP_011531415.1
XM_017004970.1 Intron XP_016860459.1
XM_017004971.1 Intron XP_016860460.1
XM_017004972.1 Intron XP_016860461.1
XM_017004973.1 Intron XP_016860462.1
XM_017004974.1 Intron XP_016860463.1
XM_017004975.1 Intron XP_016860464.1
XM_017004976.1 Intron XP_016860465.1
XM_017004977.1 Intron XP_016860466.1
XM_017004978.1 Intron XP_016860467.1
XM_017004979.1 Intron XP_016860468.1
XM_017004980.1 Intron XP_016860469.1
XM_017004981.1 Intron XP_016860470.1
XM_017004982.1 Intron XP_016860471.1
XM_017004983.1 Intron XP_016860472.1
Gene
PREPL
Gene Name
prolyl endopeptidase-like
There are no transcripts associated with this gene.

View Full Product Details