Product Details

SNP ID
rs11718920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:111048229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGTGAAATATCAGACAACTGAG[A/C]AAATAGAAATCAGTTTTGATCTCTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PVRL3-AS1 PubMed Links

Gene Details

Gene
PVRL3-AS1
Gene Name
PVRL3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details