Product Details

SNP ID
rs734107
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:41809883 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCATGCCCATCCTTGAAAATGGG[G/T]TTCCCTTCTTGTCCCCTGTTCGAAC
Phenotype
MIM: 609142
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM3 PubMed Links

Gene Details

Gene
CEACAM3
Gene Name
carcinoembryonic antigen related cell adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277163.2 Intron NP_001264092.1
NM_001815.4 Intron NP_001806.2

View Full Product Details