Product Details
- SNP ID
-
rs7012909
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:55105433 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCGCAGGCTGTCAGACAAATGTA[C/T]TAACTGACCCAGACTTGGAGAAACT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
XKR4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141955512] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- XKR4
- Gene Name
- XK related 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052898.1 |
|
Intron |
|
|
NP_443130.1 |
View Full Product Details