Product Details
- SNP ID
-
rs2251115
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:49478796 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTGCTAGGCAAACAAAACTATTC[A/G]CCATTACAATTCTAAAGGAACATAC
- Phenotype
-
MIM: 611506
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CENPQ
PubMed Links
Gene Details
- Gene
- CENPQ
- Gene Name
- centromere protein Q
View Full Product Details