Product Details

SNP ID
rs7521304
Assay Type
Validated
NCBI dbSNP Submissions
64
Location
Chr.1:160372225 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGATCCCCACCTGCGCACTATAGC[G/T]CAGGGTCAGCAGTGGAATGAAGGGC
Phenotype
MIM: 162360
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NHLH1 PubMed Links

Gene Details

Gene
NHLH1
Gene Name
nescient helix-loop-helix 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005598.3 1936 UTR 3 NP_005589.1

View Full Product Details