Product Details

SNP ID
rs4799318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31761767 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGTAGGTCACAGCATCAGACTA[C/T]ATTGCCTGCTGATGTGTACTGCCTA
Phenotype
MIM: 616153
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A52 PubMed Links

Gene Details

Gene
SLC25A52
Gene Name
solute carrier family 25 member 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034172.2 Intron NP_001029344.3

View Full Product Details