Product Details

SNP ID
rs960193
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:56318671 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTCTCTCAGCCATGTCCTACGACC[G/T]CTATGTGGCCATCTGTAACCCTCTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8K3 PubMed Links

Gene Details

Gene
OR8K3
Gene Name
olfactory receptor family 8 subfamily K member 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005202.1 365 Missense Mutation CGC,CTC R122L NP_001005202.1

View Full Product Details