Product Details

SNP ID
rs2875496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:42785845 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGTTGGATCATTTGCTGGAAAGG[C/T]GTGGGAAGCTGTCCTGTGCATTACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM216B PubMed Links

Gene Details

Gene
FAM216B
Gene Name
family with sequence similarity 216 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318932.1 Intron NP_001305861.1
NM_182508.2 Intron NP_872314.1

View Full Product Details