Product Details
- SNP ID
-
rs7917676
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:6348145 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCTTCTTGCCGCCGACTTCCTGCC[A/G]GTGCCAAGTCAGAGGGAGCTCTGCG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC399716
PubMed Links
Gene Details
- Gene
- LOC399716
- Gene Name
- uncharacterized LOC399716
There are no transcripts associated with this gene.
View Full Product Details