Product Details

SNP ID
rs3444
Assay Type
Validated
NCBI dbSNP Submissions
58
Location
Chr.1:44654293 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATGCCACAGGAATCAGCAGCCTGA[C/T]TGTTGCACTTGTCCAAACACAACTG
Phenotype
MIM: 616136
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF220 PubMed Links

Gene Details

Gene
RNF220
Gene Name
ring finger protein 220
There are no transcripts associated with this gene.

Gene
TMEM53
Gene Name
transmembrane protein 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300746.1 1446 UTR 3 NP_001287675.1
NM_001300747.1 1446 UTR 3 NP_001287676.1
NM_001300748.1 1446 UTR 3 NP_001287677.1
NM_024587.3 1446 UTR 3 NP_078863.2
XM_011542138.2 1446 UTR 3 XP_011540440.1

View Full Product Details