Product Details

SNP ID
rs11830483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15887181 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTTAAGGTATTTGGGCAATTTTT[A/T]AAAAATCTTAAACTGGCCTGTTTTG
Phenotype
MIM: 605986
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STRAP PubMed Links

Gene Details

Gene
STRAP
Gene Name
serine/threonine kinase receptor associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007178.3 Intron NP_009109.3
XM_005253294.1 Intron XP_005253351.1

View Full Product Details