Product Details

SNP ID
rs7259720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53443767 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAACCTGGATTCCATCCAAGCCAC[A/G]AGGGGTTTGTTGGGGAAAGAAAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPM3P9 PubMed Links

Gene Details

Gene
TPM3P9
Gene Name
tropomyosin 3 pseudogene 9
There are no transcripts associated with this gene.

Gene
ZNF761
Gene Name
zinc finger protein 761
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008401.3 Intron NP_001008401.3
NM_001289951.1 Intron NP_001276880.1
NM_001289952.1 Intron NP_001276881.1
NM_001289953.1 Intron NP_001276882.1

View Full Product Details